Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriphenylphosphonium bromide, 98%

Catalog No. p-7027651
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 1449-46-3 | C25H22BrP | 433.33 g/mol

Benzyltriphenylphosphonium bromide is used as a reactant for stereoselective azidolysis of vinyl epoxides, enantioselective aziridination and Friedel-Crafts cyclization for asymmetric synthesis of dihydrexidine, biomimetic iron(III) mediated oxidative dimerization for synthesis of benzoquinone parvistemin A, enantioselective synthesis of syn-diarylheptanoids from D-glucose, preparation of β-amyloid plaque ligands and decarboxylative cyclopropanation. It react with 4-methyl-oxetan-2-one to produce 4-hydroxy-1-phenyl-1-(triphenyl-l5-phosphanylidene)-pentan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyltriphenylphosphonium bromide is used as a reactant for stereoselective azidolysis of vinyl epoxides, enantioselective aziridination and Friedel-Crafts cyclization for asymmetric synthesis of dihydrexidine, biomimetic iron(III) mediated oxidative dimerization for synthesis of benzoquinone parvistemin A, enantioselective synthesis of syn-diarylheptanoids from D-glucose, preparation of β-amyloid plaque ligands and decarboxylative cyclopropanation. It react with 4-methyl-oxetan-2-one to produce 4-hydroxy-1-phenyl-1-(triphenyl-l5-phosphanylidene)-pentan-2-one.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449-46-3
Molecular Formula C25H22BrP
Molecular Weight (g/mol) 433.33
MDL Number MFCD00031707
InChI Key WTEPWWCRWNCUNA-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium bromide, bromo benzyl triphenylphosphorane, benzyltriphenylphosphanium bromide, phenylmethyl triphenylphosphonium bromide, benzyl triphenylphosphonium bromide, benzyl triphenyl phosphonium bromide, phosphonium, triphenyl phenylmethyl-, bromide, acmc-1ch0e, ksc491k3n
PubChem CID 2734970
IUPAC Name benzyl(triphenyl)phosphanium;bromide
SMILES [Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 293°C to 298°C
Quantity 10 g
UN Number UN3464
Beilstein 3599867
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 433.34
Percent Purity 98%
Chemical Name or Material Benzyltriphenylphosphonium bromide
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber 215-908-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.