Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berberine chloride

Catalog No. aaj6231106
Click to view available options
Quantity:
5 g

Catalog No. AAJ6231106

Supplier: Thermo Scientific Chemicals J6231106

Only null left
Add to Cart

CAS: 633-65-8 | C20H18ClNO4 | 371.817 g/mol

Berberine is an isoqinoline alkaloid shown to have a chemopreventive property against colon tumor formation by inhibiting the enzyme Cox-2 (cyclooxygenase-2) which is abundantly expressed in colon cancer cells. Also inhibits c-Jun (Activator Protein 1), a transcription factor that plays a critical role in inflammation and carcinogenesis. Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide (74 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), water (<1mg/mL at 25°C), and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 633-65-8
Molecular Formula C20H18ClNO4
Molecular Weight (g/mol) 371.817
MDL Number MFCD00011939
InChI Key VKJGBAJNNALVAV-UHFFFAOYSA-M
Synonym berberine chloride, berberine hydrochloride, natural yellow 18, benzodioxide, berberinium chloride, berberine hcl, unii-uot4o1byv8, berberine chloride jan, uot4o1byv8
PubChem CID 12456
ChEBI CHEBI:31271
SMILES COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.[Cl-]

Specifications

Color Yellow to Orange
Quantity 5 g
Beilstein 3836585
Solubility Information Soluble in dimethyl sulfoxide (74mg/ml at 25°C),ethanol (<1mg/ml at 25°C),water (<1mg/ml at 25°C),and methanol.
Formula Weight 371.82
Physical Form Powder
Chemical Name or Material Berberine chloride
EINECSNumber 211-195-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.