Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Berberine Monoclonal Antibody (3H10)
GREENER_CHOICE

Catalog No. PIMA549631
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549631 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549631 Supplier Invitrogen™ Supplier No. MA549631
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Berberine is a natural alkaloid compound that is commonly found in various plants, including goldenseal, barberry, and Oregon grape. It has been used in traditional medicine for centuries due to its antimicrobial, anti-inflammatory, and antioxidant properties. Berberine has also been studied for its potential health benefits, including its ability to lower blood sugar levels, reduce cholesterol, and improve gut health. It may also have potential as a treatment for certain conditions such as diabetes, metabolic syndrome, and cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Berberine
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3H10
Concentration 8.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of BBR hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.