Learn More
Siemens Healthineers Berichrom Antithrombin III (A) Reagent
Catalog No. 23044706 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
150 Tests/Kit
500 Tests/Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-706 500 Tests/Kit
23-044-707 150 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-706 Supplier Siemens Healthineers Supplier No. 10446672
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of the functional activity of antithrombin III (AT III) in human plasma.
Berichrom Antithrombin III (A) is a chromogenic activity assay for the detection of hereditary or acquired antithrombin deficiency, thrombophilia, and the monitoring of patients undergoing substitution therapy. The heparin cofactor-independent lyophilized reagent uses bovine thrombin substrate and exhibits no interference with anti-FXa anticoagulants, e.g., rivaroxaban.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Thrombophilia Testing
Quantity 500 Tests/Kit