Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BERP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583656 Supplier Invitrogen™ Supplier No. PA583656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TQRQGQEHIG SSCSFAEQAL RLGSAPEVLL VRKHMRERLA ALAAQAFPER PHENAQLELV LEVDGLRRSV LNLGALLTTS ATAHETVAT Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

The protein encoded by this gene is a member of the tripartite motif family, also called the 'RING-B-box-coiled-coil' subgroup of RING finger proteins. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic filaments. It is similar to a rat protein which is a specific partner for the tail domain of myosin V, a class of myosins which are involved in the targeted transport of organelles. The rat protein can also interact with alpha-actinin-4. Thus it is suggested that this human protein may play a role in myosin V-mediated cargo transport. Alternatively spliced transcript variants encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BERP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM3
Gene Accession No. O75382
Gene Alias Berp; BERP1; brain expressed ring finger; brain-expressed RING finger protein; Hac1; RING finger protein 22; RING finger protein 97; RING finger protein HAC1; RNF22; RNF97; Trim3; tripartite motif containing 3; tripartite motif protein 3; tripartite motif protein TRIM3; tripartite motif-containing 3; tripartite motif-containing protein 3
Gene Symbols TRIM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIM3. Recombinant protein control fragment (Product #RP-98385).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.