Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Beryllium

Catalog No. 503099348 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-9348 1 Ea.
1 options

Catalog No. 50-309-9348

Supplier: Sigma Aldrich GF522935511EA

Due to product restrictions, please to purchase or view availability for this product.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Beryllium

Specifications

CAS 7440-41-7
Melting Point 1278°C
Density 1.85 g/mL (at 25°C)
Boiling Point 2970°C
Molecular Formula Be
Linear Formula Be
Quantity 1 Ea.
MDL Number MFCD00134032
Synonym BE000240; Glucinium
Molecular Weight (g/mol) 9.01
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.