Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium AA Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical
SDP

Catalog No. p-3251355 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7440-41-7, 7697-37-2, 7732-18-5
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Be
CAS Min % 88.0, 9.91, 0.09
CAS Max % 92.0, 10.32, 0.09
Equivalence 1 mL = 1 mg Be
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Beryllium AA Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.