Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beryllium wire, 0.125mm (0.005 in.) dia., annealed, 99.7% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
25 cm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 25 cm |
| Odor | Odorless |
| Weight | ≈0.023g/m |
| Form | Wire |
| Percent Purity | 99.7% |
| Assay | (metals basis) |
| Chemical Name or Material | Beryllium wire, 0.125mm (0.005 in.) dia., Annealed, Temper: soft |
Safety and Handling
| Hazard Category | H301-H315-H317-H319-H330-H335-H350i-H372 |
| Hazard Statement | GHS H Statement H301-H330-H350-H372-H315-H319-H317-H335 Toxic if swallowed. Fatal if inhaled. May cause cancer. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c |
| EINECSNumber | 231-150-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction