Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BES, MP Biomedicals™

Catalog No. mp210092725
Click to view available options
Quantity:
25 g

Used in biochemistry and molecular biology research. BES, MP Biomedicals is a zwitterionic buffer used in the pH range of 6.15–8.35 for wide applicability. It is a sulfonic acid-containing agent that induces cross-linking between sulfonated polyimid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, N,N-Bis(2-hydroxyethyl)taurine
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Soluble 0.1M in water (clear, colorless solution)
Formula Weight 213.3
Percent Purity ≥99%
Recommended Storage Room Temperature (15–30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.