Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ Nitri-Dex™ Flock-Lined Nitrile Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.
- Ideal for jobs requiring comfort, tactility, and contact with chemical hazards
- Flocked cotton liner ensures easy donning and doffing
- Rolled cuff prevents debris from entering
- Lightweight nitrile material helps reduce hand fatigue
Specifications
Specifications
ANSI Abrasion Rating | ANSI 1 |
Certifications/Compliance | 21 CFR 170-199 |
Certifications/Compliance (Europe) | CE |
Chlorinated | No |
Color | Blue |
Latex Free | Yes |
Material | Nitrile |
Size | Small |
Thickness (English) | 9 mil |
Thickness (Metric) | 0.23 mm |
Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction