Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 707D Chemical-Protection Nitrile Gloves

Catalog No. 191501270 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-1270 Small
19-150-1274 2X-Large
19-150-1273 X-Large
19-150-1269 X-Small
19-150-1271 Medium
19-150-1272 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-150-1270 Supplier SHOWA™ Supplier No. 70707/DEL USE 19365205
This item is not returnable. View return policy

The 707D chemical-protection gloves combine the best of disposable and chemical-resistant technology. This hybrid results in the ideal solution for jobs requiring contact with chemical hazards, optimum comfort, and tactility.

  • Form-fitting
  • Easy donning and doffing
  • Chemical-resistant
  • Oil-resistant
  • Water-resistant
  • Abrasion-resistant
  • Hydrocarbon-resistant
  • Impermeable
  • Biodegradable
  • Ergonomic
  • Rolled Cuff
  • 100% Nitrile
  • Chlorinated
  • Embossed grip
  • Unlined
  • Eco Best Technology™ (EBT)
  • Industries: Chemical, Food, Janitorial, Laboratory, Mining, Pharmaceutical

Specifications

Certifications/Compliance 21 CFR 170-199, ANSI/ISEA 105-2005
Color Blue
Latex Free Yes
Material Nitrile
Size Small
Thickness (English) 9 mil
For Use With (Application) manufacturing, food processing, laboratories, janitorial, paint applications, poultry processing, light duty maintenance and clean up
Glove Cuff Style Rolled
Grip Design Rough
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.