Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Zorb-IT Extra Work Gloves

Catalog No. 19168931 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
7
8
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-931 7
19-168-932 8
19-168-933 9
19-168-934 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-168-931 Supplier SHOWA™ Supplier No. 457507
This item is not returnable. View return policy

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile
Color Gray, White
Exterior Finish No Texture
Material Nylon
Product Type Dip-coated Gloves
Size 7
For Use With (Application) Material Handling, Metal Fabrication, and Assembly
Glove Cuff Style Elasticated
Series 4575
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.