Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Zorb-IT Ultra Work Gloves

Catalog No. 19172104 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-172-104 11
19-172-100 7
19-172-101 8
19-172-102 9
19-172-103 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-172-104 Supplier SHOWA™ Supplier No. 456511
This item is not returnable. View return policy

Work gloves ideal for medium to heavy duty tasks in a variety of industries.

Specifications

Color Gray, Yellow
Cut Resistance Rating ANSI A3
Material Aramid Fiber, Lycra
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Automotive, construction, glass, metallurgy
Glove Cuff Style Elasticated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.