Learn More
Description
- 100% nitrile, latex-free
- Powder free
- Increased dexterity
- 4 mil thick, 9.5 in. length
- Protects hands from an array of chemical hazards
- Rolled cuff prevents debris from entering the glove
- Second-skin feel reduces hand fatigue
- Ideal for a variety of applications including blood drawing, dental hygiene, food preparation, EMS, lab work and pharmaceutical
- Made in the USA
Compliance
Data Testing is available upon request.
Specifications
Specifications
| Certifications/Compliance | 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 |
| Applications | Laboratory, Healthcare, Industrial |
| AQL | 1.5 |
| Color | Blue |
| Disposable | Single-use |
| Exterior Finish | No Texture |
| Latex Free | Yes |
| Material | Nitrile |
| Powdered | Powder Free |
| Size | Medium |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
