Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ N-DEX™ 7005 Nitrile Gloves

Catalog No. 19050269D Shop All Showa Best Glove Products
Change view
Click to view available options
Certifications/Compliance:
21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526
Size:
X-Small
Small
Medium
Large
X-Large
Thickness (English):
4 mil
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Size Thickness (English)
19-050-269D 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 X-Large 4 mil
19-050-269C 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 Large 4 mil
19-050-269AA 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 X-Small 4 mil
19-050-269A 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 Small 4 mil
19-050-269B 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526 Medium 4 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-050-269D Supplier SHOWA™ Supplier No. 7005PFXL
This item is not returnable. View return policy

Nitrile Gloves provide unmatched dexterity and performance across a range of tactical applications.

  • 100% nitrile, latex-free
  • Powder free
  • Increased dexterity
  • 4 mil thick, 9.5 in. length
  • Protects hands from an array of chemical hazards
    • Rolled cuff prevents debris from entering the glove
    • Second-skin feel reduces hand fatigue
  • Ideal for a variety of applications including blood drawing, dental hygiene, food preparation, EMS, lab work and pharmaceutical
  • Made in the USA

Compliance

Data Testing is available upon request.

Specifications

Certifications/Compliance 21 CFR 170-199, FDA, NFPA, OSHA, CFIA, ASTM D5526
Applications Laboratory, Healthcare, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 7005PFXL
Tensile Strength 14 MPa
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.