Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Heavy-Duty Chemical Resistant Latex Gloves

Catalog No. 19019882 Shop All Showa Best Glove Products
Change view
Click to view available options
15in. Long (38.1cm); Size 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-019882 15in. Long (38.1cm); Size 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-019882 Supplier SHOWA™ Supplier No. 5509
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Chemical resistant glove is designed for handling hazardous acids and other volatile substances. Two-colored construction serves as a wear-indicator for added protection and resistance.

Heavy-duty gloves are ideal for dip tank, sandblasting, and electroplating applications

  • Acid- and abrasion-resistant exterior features
  • Bisque finish for secure gripping
  • Inside of gloves is smooth and unflocked
  • Powdered interior and rolled-edge cuffs expedite donning and doffing
  • Two-color composition (black exterior/red interior) serves as a visual wear indicator
  • Hand specific, non-sterile, and liquid proof

Gloves are not intended for electrical use

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 388 1120, CE
Color Black, Red
Material Latex
Size Large
Thickness (English) 40 mil
Thickness (Metric) 1 mm
EN388 Standard EN 388:2011X
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES AKLMT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Manufacturing, aircraft assembly, janitorial, plant maintenance, agriculture, heavy duty plant maintenance, chemical processing, sanitization clean up, sandblasting/electroplating
Glove Cuff Style Rolled
Grip Design Bisque
Lining Unlined
Model 55-09
Length (English) 15 in.
Length (Metric) 38.1 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.