Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Nitri-Flex™ Lite™ Nitrile Coated Chemical Resistant Gloves

Catalog No. 19821994 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
7
8
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-994 8
19-821-993 7
19-821-996 10
19-821-995 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-821-994 Supplier SHOWA™ Supplier No. G4500S08
This item is not returnable. View return policy

Flat-dipped gloves provide tactile dexterity and comfort

  • Ideal for jobs requiring tactile discrimination plus puncture and abrasion protection
  • Gloves deliver a lower electrostatic discharge than comparable latex-coated gloves
  • Nitrile-dipped palm

Specifications

ANSI Abrasion Rating ANSI 1
Coated Area Palm
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Small Parts Assembly, Inspection, Fabrication, Material Handling, and Shipping/Receiving
Glove Cuff Style Knit
Series G4500S
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.