Learn More
BEST3 Mouse anti-Human, Clone: OTI3B11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501768

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BEST3 belongs to the bestrophin family of anion channels, which includes BEST1, the gene mutant in vitelliform macular dystrophy (VMD), and 2 other BEST1-like genes, BEST2 and BEST4. Bestrophins are transmembrane (TM) proteins that share a homology region containing a high content of aromatic residues, including an invariant arg-phe-pro (RFP) motif. The bestrophin genes share a conserved gene structure, with almost identical sizes of the 8 RFP-TM domain-encoding exons and highly conserved exon-intron boundaries. Each of the 4 bestrophin genes has a unique 3-prime end of variable length.Specifications
BEST3 | |
Monoclonal | |
Unconjugated | |
BEST3 | |
VMD2L3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
144453 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3B11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8N1M1 | |
BEST3 | |
Full length human recombit protein of human BEST3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.