Learn More
Bestrophin 1 Monoclonal Antibody (E6-6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA191895
Description
MA1-91895 detects Bestrophin from non-human primate, human samples. MA1-91895 has been successfully used in immunofluorescence, Western blot applications. The MA1-91895 immunogen is: Partial recombit protein [KDHMDPYWALENRDEAHS] Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Best macular dystrophy (BMD) or vitelliform macular dystrophy (VMD2), is an autosomal form of macular degeneration, characterized by a depressed light peak in the electrooculogram (EOG). It is inherited and has an early onset. Bestrophin is a 68 kDa bas
Specifications
Bestrophin 1 | |
Monoclonal | |
Unconjugated | |
O76090 | |
BEST1 | |
Partial recombit protein [KDHMDPYWALENRDEAHS] | |
RUO | |
7439 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
E6-6 | |
BEST1 | |
ARB, BEST, BMD, RP50, TU15B, VMD2 | |
Mouse | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.