Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bestrophin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111228 Supplier Invitrogen™ Supplier No. PA5111228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRLSFLLRKN SCVSEASTGA SCSCAVVPEG AAPECSCGDP LLDPGLPEPE APPPAGPEPL TLIPGPVEPF SIVTMP Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 65%.

Best2 forms calcium-sensitive chloride channels and is permeable to bicarbonate. The Best2 gene is a member of the bestrophin gene family of anion channels. Bestrophin genes share a similar gene structure with highly conserved exon-intron boundaries, but with distinct 3' ends. Bestrophins are transmembrane proteins that contain a homologous region rich in aromatic residues, including an invariant arg-phe-pro motif. Mutation in the Best2 gene is associated with vitelliform macular dystrophy. The Best2 gene is mainly expressed in the retinal pigment epithelium and colon. Diseases associated with BEST2 include Vitelliform Macular Dystrophy and Bestrophinopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bestrophin 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BEST2
Gene Accession No. Q8NFU1
Gene Alias BEST2; bestrophin 2; bestrophin-2; Hbest2; vitelliform macular dystrophy 2-like 1; vitelliform macular dystrophy 2-like protein 1; VMD2L1; VMD2L-1; VMD2-like gene 1
Gene Symbols BEST2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Bestrophin 2. Recombinant protein control fragment (Product #RP-98318).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54831
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.