Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114900 Supplier Thermo Scientific Supplier No. PA5114900
Add to Cart
Add to Cart

Bestrophin 2 Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bestrophin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene BEST2
Gene Accession No. Q8BGM5, Q8NFU1
Gene Alias BEST2; bestrophin 2; bestrophin-2; Hbest2; vitelliform macular dystrophy 2-like 1; vitelliform macular dystrophy 2-like protein 1; VMD2L1; VMD2L-1; VMD2-like gene 1
Gene Symbols BEST2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BEST2(Accession Q8NFU1), corresponding to amino acid residues T429-E479.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212989, 364973, 54831
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.