Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bestrophin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533365
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Bat (95%) Hamster, Elephant, Panda, Bovine, Pig (90%) Dog, Rabbit (85%).
BEST3 belongs to the bestrophin family of anion channels, which includes BEST1, the gene mutant in vitelliform macular dystrophy (VMD), and 2 other BEST1-like genes, BEST2 and BEST4. Bestrophins are transmembrane (TM) proteins that share a homology region containing a high content of aromatic residues, including an invariant arg-phe-pro (RFP) motif. The bestrophin genes share a conserved gene structure, with almost identical sizes of the 8 RFP-TM domain-encoding exons and highly conserved exon-intron boundaries. Each of the 4 bestrophin genes has a unique 3-prime end of variable length.
Specifications
Bestrophin 3 | |
Polyclonal | |
Unconjugated | |
BEST3 | |
BEST3; bestrophin 3; bestrophin-3; mBest4; vitelliform macular dystrophy 2-like 3; vitelliform macular dystrophy 2-like protein 3; Vmd2l3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
144453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8N1M1 | |
BEST3 | |
Synthetic 20 amino acid peptide from internal region of human BEST3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction