Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bestrophin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533367
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533367 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533367 Supplier Invitrogen™ Supplier No. PA533367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Bat, Rabbit, Pig (100%) Marmoset, Rat, Horse (94%) Dog (89%) Panda, Lizard (83%).

This gene is a member of the bestrophin gene family of anion channels. Bestrophin genes share a similar gene structure with highly conserved exon-intron boundaries, but with distinct 3' ends. Bestrophins are transmembrane proteins that contain a homologous region rich in aromatic residues, including an invariant arg-phe-pro motif. Mutation in one of the family members (bestrophin 1) is associated with vitelliform macular dystrophy. The bestrophin 4 gene is predominantly expressed in the colon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bestrophin 4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene BEST4
Gene Accession No. Q8NFU0
Gene Alias BEST4; bestrophin 4; bestrophin-4; si:ch211-106k21.4; similar to vitelliform macular dystrophy 2-like 2; vitelliform macular dystrophy 2-like 2; vitelliform macular dystrophy 2-like protein 2; VMD2L2
Gene Symbols BEST4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human BEST4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100345949, 100516679, 266675, 510542
Target Species Human, Rabbit, Bovine, Pig, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.