Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502206214
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6214 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6214 Supplier Fabgennix Inc Supplier No. BEST121AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bestrophin Polyclonal antibody specifically detects Bestrophin in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Bestrophin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene BEST1
Gene Accession No. O76090
Gene Alias ARB; BEST; BEST 1; Best disease; best macular dystrophy; BEST1; Best1V1Delta2; bestrophin; bestrophin 1; Bestrophin1; Bestrophin-1; BMD; Bmd1; LOW QUALITY PROTEIN: bestrophin-1; mBest1; RP50; TU15B; vitelliform macular dystrophy (Best disease, bestrophin); vitelliform macular dystrophy 2 homolog; vitelliform macular dystrophy protein 2; Vitelliform macular dystrophy protein 2 homolog; VMD 2; Vmd2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 500-585 on human Bestrophin-1 isoform 1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7439
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.