Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BET1L Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305961PE
Description
BET1L Polyclonal antibody specifically detects BET1L in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
BET1L | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
blocked early in transport 1 homolog (S. cerevisiae)-like, golgi integral membrane protein 3, golgi SNARE 15 kDa protein, golgi SNARE with a size of 15 kDa, GS15, HSPC197, vesicle transport protein GOS15 | |
Produced in rabbits immunized with E. coli-derived Human BET1L fragment. | |
0.1 mL | |
Signal Transduction | |
51272 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction