Learn More
Invitrogen™ BET1L Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | BET1L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Bet1l |
| Gene Accession No. | Q9NYM9 |
| Gene Alias | 2610021K23Rik; Bet1; Bet1 golgi vesicular membrane trafficking protein like; Bet1 golgi vesicular membrane trafficking protein-like; Bet1l; bet1l {ECO:0000312; BET1L1; BET1-like protein; blocked early in transport 1 homolog (S. cerevisiae)-like; blocked early in transport 1 homolog like; blocked early in transport 1 homolog-like; golgi integral membrane protein 3; golgi SNARE 15 kDa protein; golgi SNARE with a size of 15 kDa; golgi SNARE, 15 kD; Golgi soluble N-ethylmaleimide-sensitive factor attachment protein 15; GOLIM3; GOS-15; Gs15; HSPC197; RGD:71051}; soluble N-ethylmaleimide-sensitive factor attachment protein (SNARE) 15; vesicle transport protein GOS15 |
| Gene Symbols | Bet1l |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.