Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BET1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729256 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9256 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9256 Supplier Proteintech Group Inc Supplier No. 141631AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vesicle SNARE required for targeting and fusion of retrograde transport vesicles with the Golgi complex. Required for the integrity of the Golgi complex.

Specifications

Antigen BET1L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BET1L
Gene Accession No. O35152, O35153, Q9NYM9
Gene Alias BET1 like protein, BET1L, BET1L1, GOLIM3, GOS 15, GS15, HSPC197
Gene Symbols BET1L
Host Species Rabbit
Immunogen BET1L Fusion Protein Ag5351
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51272, 54399, 54400
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.