Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ beta-1,2 Adaptin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141069
This item is not returnable.
View return policy
Description
beta-1,2 Adaptin Polyclonal Antibody for Western Blot, IHC (P), Flow
Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as one of the large subunits of this complex and is a member of the adaptin protein family. This gene is a candidate meningioma gene. Alternative splicing results in multiple transcript variants.
Specifications
| beta-1,2 Adaptin | |
| Polyclonal | |
| Unconjugated | |
| AP1B1 | |
| adapter-related protein complex 1 subunit beta-1; adapter-related protein complex 2 beta subunit; adapter-related protein complex 2 subunit beta; adaptin, beta 2 (beta); adaptor protein complex AP-1 subunit beta-1; adaptor protein complex AP-1, beta 1 subunit; adaptor protein complex AP-2 subunit beta; adaptor related protein complex 1 beta 1 subunit; adaptor related protein complex 1 subunit beta 1; adaptor related protein complex 2 beta 1 subunit; adaptor related protein complex 2 subunit beta 1; adaptor related protein complex 2, beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; adaptor-related protein complex 2 subunit beta; adaptor-related protein complex 2, beta 1 subunit; adaptor-related protein complex AP-1 beta 1 subunit; ADTB1; ADTB2; AP-1 complex subunit beta-1; AP105A; AP105B; AP1B1; AP-2 complex subunit beta; AP2B1; AP2-BETA; BAM22; beta adaptin; beta-1-adaptin; beta1-adaptin; Beta-2-adaptin; beta2-adaptin; beta-adaptin; Beta-adaptin 1; beta-prime-adaptin; Clapb1; CLAPB2; Clathrin assembly protein complex 1 beta large chain; clathrin assembly protein complex 2 beta large chain; clathrin-associated / assembly / adaptor protein, large, beta 1; clathrin-associated/assembly/adaptor protein, large, beta 1; golgi adaptor HA1/AP1 adaptin beta subunit; MGC128441 protein; plasma membrane adaptor HA2/AP2 adaptin beta subunit; plasma membrane adaptor HA2/AP2 adaptor beta subunit; testicular tissue protein Li 22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11764, 140670, 162, 163, 29663, 71770 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O35643, P52303, P62944, P63010, Q10567, Q9DBG3 | |
| AP1B1, AP2B1 | |
| Recombinant protein within human AP2B1 aa 400-600/937. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction