Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (3B8C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
---|---|
Clone | 3B8C12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61709
![]() |
Novus Biologicals
NBP261709 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse | |
27087 | |
Purified recombinant fragment of human B3GAT1 (AA: 193-334) expressed in E. Coli. | |
Primary | |
38.2 kDa |
3B8C12 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
Monoclonal | |
Purified | |
Cancer, Immunology | |
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
B3GAT1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title