Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-1,3-Glucuronyltransferase 1/B3GAT1 Antibody (CL9499), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32117025UL
Description
beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects beta-1,3-Glucuronyltransferase 1/B3GAT1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
CL9499 | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
This antibody was developed against a Synthetic Peptide of Q9P2W7. Actual immunogen sequence is proprietary. | |
25 μg | |
Cancer, Immunology | |
27087 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction