Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-1,3-Glucuronyltransferase 3/B3GAT3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299973JF646
Description
beta-1,3-Glucuronyltransferase 3/B3GAT3 Polyclonal specifically detects beta-1,3-Glucuronyltransferase 3/B3GAT3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
beta-1,3-Glucuronyltransferase 3/B3GAT3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Beta-1,3-glucuronyltransferase 3, beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3, GlcAT-I, GLCATI, GlcUAT-I, Glucuronosyltransferase I, Sqv-8-like protein, UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase | |
Produced in rabbits immunized with purified, recombinant beta-1,3-Glucuronyltransferase 3/B3GAT3 (NP_036332.2; Glu72-Val335). | |
0.1 mL | |
metabolism | |
26229 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction