Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 Polyclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
11041 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EEQDQYFEFFPPSPRSVDQVKAQLRTALASGGVLDASGDYRVYRGLLKTTMDPNDVILATHASVDNLLHLSGLLERWEGPLSVSVFAATKEEAQLATVLAYALSSHCPDMRARVAMHLVCPSRYEAAVPDPREPGEFALLRSCQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
B3GN-T1, beta-1,3-N-acetylglucosaminyltransferase bGnT-6, BETA3GNTI, EC 2.4.1.149, i-beta-1,3-N-acetylglucosaminyltransferase, iGAT, IGNT, N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase, Poly-N-acetyllactosamine extension enzyme, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1B3GNT6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | |
B4GAT1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title