Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 Polyclonal antibody specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Beta-1,3-N-Acetylglucosaminyltransferase 6/B3GNT6 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Endocrinology, Signal Transduction | |
PBS (pH 7.2), 40% Glycerol | |
192134 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
B3Gn-T6, beta-1,3-Gn-T6, Beta-1,3-N-acetylglucosaminyltransferase 6, beta-1,3-N-acetylglucosaminyltransferase protein, beta3Gn-T6, BGnT-6, Core 3 synthase, EC 2.4.1.-, IMAGE:4907098, MGC119334, MGC119336, MGC119337, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase) | |
This antibody was developed against a recombinant protein corresponding to amino acids: QQWFLQAPRSPREERSPQEETPEGPTDA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title