Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta-1 Adaptin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566752 Supplier Invitrogen™ Supplier No. PA566752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPSAFVEGGRG VVHKSLPPRT ASSESAESPE TAPTGAPP Highest antigen sequence identity to the following orthologs - mouse 90%, rat 87%.

AP-1 complex subunit beta-1 is a subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-1 Adaptin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ap1b1
Gene Accession No. Q10567
Gene Alias adapter-related protein complex 1 subunit beta-1; adaptor protein complex AP-1 subunit beta-1; adaptor protein complex AP-1, beta 1 subunit; adaptor related protein complex 1 beta 1 subunit; adaptor-related protein complex 1 beta 1 subunit; adaptor-related protein complex 1 subunit beta-1; adaptor-related protein complex 1, beta 1 subunit; adaptor-related protein complex AP-1 beta 1 subunit; adaptor-related protein complex AP-1, beta 1 subunit; ADTB1; ADTB2; AP-1 complex subunit beta-1; AP105A; AP105B; Ap1b1; AP2B1; b2b1660Clo; BAM22; Beta-1-adaptin; beta1-adaptin; Beta-2-adaptin; Beta-adaptin; beta-adaptin 1; beta-prime adaptin; beta-prime-adaptin; CLAPB1; CLAPB2; Clathrin assembly protein complex 1 beta large chain; golgi adaptor HA1/AP1 adaptin beta subunit; Mutant line 1660; plasma membrane adaptor HA2/AP2 adaptor beta subunit
Gene Symbols Ap1b1
Host Species Rabbit
Immunogen Recombinant Human beta-1 Adaptin. Recombinant protein control fragment (Product #RP-105349).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.