Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Antibody (4C11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | beta-2 Adrenergic R/ADRB2 |
---|---|
Clone | 4C11 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
beta-2 Adrenergic R/ADRB2 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
A synthesized peptide derived from Human beta-2 Adrenergic R/ADRB2 [UniProt P07550] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
4C11 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
154 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title