Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-2 Adrenergic R/ADRB2 Mouse anti-Human, Clone: 13D6, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | beta-2 Adrenergic R/ADRB2 |
---|---|
Clone | 13D6 |
Concentration | 1 mg/ml |
Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
Description
beta-2 Adrenergic R/ADRB2 Monoclonal antibody specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunoprecipitationSpecifications
beta-2 Adrenergic R/ADRB2 | |
1 mg/ml | |
Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction | |
PBS | |
154 | |
IgG1 κ | |
Protein A purified |
13D6 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor | |
This antibody was raised by immunizing mice with beta-2 adrenergic receptor. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title