Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-2 Adrenergic R/ADRB2 Antibody (4A6C9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261711 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61711 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61711 Supplier Novus Biologicals Supplier No. NBP261711
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4A6C9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:100000
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADRBR, adrenergic, beta-2-, receptor, surface, B2ARADRB2R, BAR, beta-2 adrenergic receptor, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Gene Symbols ADRB2
Host Species Mouse
Immunogen Purified recombinant fragment of human ADRB2 (AA: 302-413) expressed in E. Coli.
Molecular Weight of Antigen 65.4 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Asthma, GPCR, Immunology, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 154
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.