Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ beta-2 Adrenergic Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533334
Description
beta-2 Adrenergic Receptor Polyclonal Antibody for ICC/IF, IHC (P)
Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence.
Specifications
| beta-2 Adrenergic Receptor | |
| Polyclonal | |
| Unconjugated | |
| Adrb2 | |
| Adrb2; Adrb-2; ADRB2R; ADRBR; Adrenergic beta 2- receptor surface; adrenergic receptor beta 2; adrenergic receptor, beta 2; adrenergic, beta-2, receptor, surface; adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; AR beta2; B2AR; Badm; BAR; beta 2-AR; beta-2 adrenergic receptor; Beta-2 adrenoceptor; beta-2 adrenoreceptor; beta-2-adrenergic receptor; beta2-adrenergic receptor; beta2-Adrenoreceptor; BETA2AR; catecholamine receptor; Gpcr7; HGNC:286 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 154 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P07550 | |
| Adrb2 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human ADRB2. Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey, Marmoset (88%); Mouse (81%). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction