Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech beta-2 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P161G50
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P161G50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 500P161G50 Supplier PeproTech Supplier No. 500P161G50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: GIGDPVTCLK SGAICHPVFC PRRYKQIGTC GLPGTKCCKK P. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human BD-2. Anti-Human BD-2-specific antibody was purified by affinity chromatography employing an immobilized Human BD-2 matrix. Sandwich ELISA: To detect Human BD-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-2 (500-P161GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-2. Western Blot: To detect Human BD-2 by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Defensin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DEFB4A
Gene Accession No. O15263
Gene Alias BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4A; Defensin beta2; defensin, beta 2; defensin, beta 4; defensin, beta 4A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1
Gene Symbols DEFB4A
Host Species Goat
Immunogen E.coli-derived Recombinant Human BD-2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.