Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta 2 Defensin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986052
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6052 Supplier Bioss Supplier No. BS4307R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibiotic peptide is locally regulated by inflammation. Defensins form a family of microbicidal and cytotoxic peptides made by neutrophilis. Members of the defensin family are highly similar in protein sequence.

Specifications

Antigen beta 2 Defensin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DEFB4A
Gene Accession No. O15263
Gene Alias BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4A; Defensin beta2; Defensin, beta 2; defensin, beta 4; defensin, beta 4A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1
Gene Symbols DEFB4A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human beta 2 Defensin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.