Learn More
Beta-2-Glycoprotein 1 Mouse anti-Human, Clone: 7B8, Bethyl Laboratories
Mouse Monoclonal Antibody
Supplier: Bethyl Laboratories, Inc A500007AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.Specifications
Beta-2-Glycoprotein 1 | |
Monoclonal | |
Unconjugated | |
APOH | |
activated protein C-binding protein, anticardiolipin cofactor, APC inhibitor, apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, beta(2)GPI, beta-2-glycoprotein I, BG | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
350 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
7B8 | |
TBS with 0.09% sodium azide | |
P02749 | |
APOH | |
Purified Beta-2-Glycoprotein from normal human serum | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.