Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247700PECY7
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry.Specifications
beta 2-Microglobulin | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
B2M | |
Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient. | |
0.1 mL | |
Primary | |
Recognizes a protein of 12kDa, identified as beta-microglobulin. Beta--microglobulin non-covalently associates with the 44kDa chain to form the HLA Class I antigen complex. Human beta-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This monoclonal antibody is specific to human beta-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. Detection of beta-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
246-E9.E7 (same as HLA.ABC.m2) | |
Flow Cytometry | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
567.0 | |
Human, Primate (Negative) | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction