Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Antibody (BBM.1), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281045H
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
beta 2-Microglobulin | |
Monoclonal | |
HRP | |
PBS | |
B2M | |
Molt 4, a human T cell line. | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
567 | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
BBM.1 | |
Western Blot, Immunohistochemistry | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Binds human beta 2-Microglobulin, a component of MHC class I molecules, which are present on all nucleated cells (excludes red blood cells) and involved in the presentation of peptide antigens to the immune system. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction