Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Antibody (C21.48A1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311404R
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
beta 2-Microglobulin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
C21.48A1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Soluble beta 2-Microglobulin (Uniprot: P61769) | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
567 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction