Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-2 Microglobulin/FCRN Polyclonal Antibody
beta-2 Microglobulin/FCRN Polyclonal Antibody
Supplier: Thermo Scientific PA580367
Description
beta-2 Microglobulin/FCRN Polyclonal Antibody for IHC (P)

Specifications
beta-2 Microglobulin/FCRN | |
Polyclonal | |
Unconjugated | |
FCGRT | |
alpha-chain; B2M; beta chain of MHC class I molecules; beta-2 microglobulin; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; CDABP0092; Fc fragment immunoglobulin G receptor; Fc fragment of IgG receptor and transporter; Fc fragment of IgG, receptor, transporter, alpha; Fc receptor, IgG, alpha chain transporter; FCGRT; FCRN; FcRn alpha chain; HDCMA22P; heavy chain of the major histocompatibility complex class I-like Fc receptor; igG Fc fragment receptor transporter alpha chain; IgG receptor FcRn large subunit p51; IMD43; immunoglobulin receptor, intestinal, heavy chain; major histocompatibility complex class I-like Fc receptor; neonatal Fc receptor; neonatal Fc-receptor for Ig; transmembrane alpha chain of the neonatal receptor | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2217, 567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
P55899, P61769 | |
B2m, FCGRT | |
Recombinant Human FCGRT & B2M Heterodimer Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction