Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta-2 Microglobulin Monoclonal Antibody (9-E10-C6)
GREENER_CHOICE

Catalog No. KSK0030102
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KSK0030102 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KSK0030102 Supplier Invitrogen™ Supplier No. KSK0030102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KSK 003-01-02 reacts with free as well as MHC class I associated b2-microglobulin from the genus Caiman. KSK 003-01-02 has been successfully used in immunohistochemistry and Western blotting techniques. KSK 003-01-02 is suitable for Western blotting, flow cytometry, immunoprecipitation, and affinity purification on caiman b2-microglobulin. KSK 003-01-02 reacts with a non-polymorphic epitope on caiman b2-microglobulin. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Beta-2-microglobulin (B2M) is a 12-14 kDa protein that is a subunit of the major histocompatibility complex (MHC) class I. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells, but is absent on erythrocytes. Beta-2 microglobulin is also found in free form in the plasma. The amino acid sequence of Beta-2-microglobulin is homologous to the constant regions of immunoglobulin heavy and light chains, with a predominantly beta pleated sheet structure. The protein is ubiquitously expressed in the cytoplasm, and is involved in antigen processing and presentation. Beta-2-microglobulin also plays a role in TCR signaling, and downstream signaling in naive CD8+ cells. Beta 2-microglobulin is encoded by the B2M gene located on chromosome 15. A mutation in the Beta-2-microglobulin gene has been shown to result in hypercatabolic hypoproteinemia. Levels of beta 2 microglobulin can be used to assess renal function in kidney transplant recipients. Beta-2-microglobulin that protein can be used as a prognostic marker for HIV infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Microglobulin
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9-E10-C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Alias B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m
Host Species Mouse
Immunogen Recombinant Caiman beta2-microglobulin.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Reptile
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.