Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Mouse anti-Rat, Clone: TLD-3H12B, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB60013830.025MG
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P07151 | |
| B2M | |
| Rat microglial cells | |
| 0.025 mg | |
| Adaptive Immunity, Cancer, Diabetes Research, Immunology | |
| 567 | |
| Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry | |
| TLD-3H12B | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65517 recognizes rat beta-2 microglobulin, expressed at the surface of most cells in association with MHC Class I molecules. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction