Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta 2-Microglobulin Mouse anti-Rat, Clone: TLD-3H12B, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001383
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
beta 2-Microglobulin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-65517 recognizes rat beta-2 microglobulin, expressed at the surface of most cells in association with MHC Class I molecules. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
TLD-3H12B | |
Unconjugated | |
P07151 | |
B2M | |
Rat microglial cells. | |
0.125 mg | |
Adaptive Immunity, Cancer, Diabetes Research, Immunology | |
567 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction