Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta-2 Microglobulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142897
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142897 Supplier Invitrogen™ Supplier No. PA5142897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Beta-2-microglobulin (B2M) is a 12-14 kDa protein that is a subunit of the major histocompatibility complex (MHC) class I. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells, but is absent on erythrocytes. Beta-2 microglobulin is also found in free form in the plasma. The amino acid sequence of Beta-2-microglobulin is homologous to the constant regions of immunoglobulin heavy and light chains, with a predominantly beta pleated sheet structure. The protein is ubiquitously expressed in the cytoplasm, and is involved in antigen processing and presentation. Beta-2-microglobulin also plays a role in TCR signaling, and downstream signaling in naive CD8+ cells. Beta 2-microglobulin is encoded by the B2M gene located on chromosome 15. A mutation in the Beta-2-microglobulin gene has been shown to result in hypercatabolic hypoproteinemia. Levels of beta 2 microglobulin can be used to assess renal function in kidney transplant recipients. Beta-2-microglobulin that protein can be used as a prognostic marker for HIV infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Microglobulin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene B2m
Gene Accession No. P61769
Gene Alias B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m
Gene Symbols B2m
Host Species Goat
Immunogen Peptide with sequence C-RIEKVEHSDLSFSKD (aa 65-79).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 567
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.