Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-3 Adrenergic R/ADRB3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215567
Description
beta-3 Adrenergic R/ADRB3 Polyclonal specifically detects beta-3 Adrenergic R/ADRB3 in Human samples. It is validated for Western Blot.Specifications
beta-3 Adrenergic R/ADRB3 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ADRB3R, adrenergic, beta-3-, receptor, B3AR, beta-3 adrenergic receptor, Beta-3 adrenoceptor, Beta-3 adrenoreceptor, BETA3AR, FLJ99960 | |
Rabbit | |
44 kDa | |
0.1 mL | |
GPCR, Neuroscience | |
155 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.82mg/mL | |
Western Blot 1:500-1:3000 | |
P13945 | |
ADRB3 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human beta 3 adrenergic receptor. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction